CPC® MC Quick-Disconnect Fittings, Threaded Body & Insert, Non-Valved, Chrome-Plated Brass
Provides quick and secure connection and disconnection of fluid lines
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly, even with gloved hands
  • Withstands higher pressure and temperature
  • Mix and match termination sizes for added flexibility
  • Compatible with PMC acetal and PMC12 polypropylene couplings
The CPC (Colder) MC series quick-disconnect threaded body and insert fittings provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. An ergonomic thumb latch allows you to safely connect and disconnect the fittings with the use of one hand, even with gloves on.
These chrome-plated brass fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures.
1,500.00 - ₹2,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) MC Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Non-Valved, 1/8" NPT; 1/Each 5013571 Mfr # MC1002
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valve 1/8
₹2,600.00INR / Each
CPC (Colder) MC Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Non-Valved, 1/4" NPT; 1/Each 5013572 Mfr # MC1004
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valve 1/4
₹2,700.00INR / Each
CPC (Colder) MC Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Non-Valved, 1/8" NPT; 1/Each 5013578 Mfr # MC2402
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valve 1/8
₹1,500.00INR / Each
CPC (Colder) MC Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Non-Valved, 1/4" NPT; 1/Each 5013579 Mfr # MC2404
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valve 1/4
₹1,600.00INR / Each