CPC® (Colder) Steam-Thru® Sanitary Steam-in-Place Connectors, Single Cycle, Polysulfone
Make a quick and easy sterile connection between stainless steel processing equipment and tubing assemblies
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Innovative three-port steam design
  • Patented steam valve design
  • 3/4" sanitary connection for condensate port
  • 0.5" nominal flow path size
The CPC® (Colder) Steam-Thru® connectors feature a unique three-port design for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. Use the Steam-Thru SIP connection to sterilize the fitting while connected to stainless steel equipment. Steam passes directly through the lower port to stainless steel equipment, then once the SIP cycle is completed, the valve is actuated to create a sterile flow path to single-use systems. Select from 3/4" and 1-1/2" sanitary connections for attachment to the processing equipment. Connectors withstand temperatures up to 130°C (266°F) for 60 minutes during SIP.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
27,000.00 - ₹27,300.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Sanitary Clamp Size (Port 2) (in) Sanitary Clamp Size (Port 3) (InchShort)
Availability Pricing
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Each 5011341 Mfr # STC1700000
COMPARE
1/2 3/4 3/4
₹27,000.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 3/8" ID x 3/4" x 3/4" Sanitary; 1/Each 5011342 Mfr # STC1700100
COMPARE
3/8 3/4 3/4
₹27,200.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 1/2" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011343 Mfr # STC1700200
COMPARE
1/2 3/4 1-1/2
₹27,000.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 3/8" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011344 Mfr # STC1700300
COMPARE
3/8 3/4 1-1/2
₹27,200.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Each 5011345 Mfr # STC1700500
COMPARE
1/2 3/4 3/4
₹27,300.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polycarbonate Sleeve, 3/8" ID x 3/4" x 3/4" Sanitary; 1/Each 5011346 Mfr # STC1700600
COMPARE
3/8 3/4 3/4
₹27,000.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011347 Mfr # STC1700700
COMPARE
1/2 3/4 1-1/2
₹27,200.00INR / Each
CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polycarbonate Sleeve, 3/8" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011348 Mfr # STC1700800
COMPARE
3/8 3/4 1-1/2
₹27,000.00INR / Each
Showing 1- 8 of 8