CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Each

Representative image only.

CPC (Colder) Steam-Thru® Sanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Each

CPC (Colder) | Mfr: STC1700000| Item: 5011341

Make a quick and easy sterile connection between stainless steel processing equipment and tubing assemblies

₹27,000.00INR / Each

₹27,000.00INR / Each
Product Description

Key Features

  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Innovative three-port steam design
  • Patented steam valve design
  • 3/4" sanitary connection for condensate port
  • 0.5" nominal flow path size
  • Polysulfone with polyethylene sleeve
  • 1/2" hose barb x 3/4" sanitary connection

More About this Item

The CPC® (Colder) Steam-Thru® connectors feature a unique three-port design for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. Use the Steam-Thru SIP connection to sterilize the fitting while connected to stainless steel equipment. Steam passes directly through the lower port to stainless steel equipment, then once the SIP cycle is completed, the valve is actuated to create a sterile flow path to single-use systems. Select from 3/4" and 1-1/2" sanitary connections for attachment to the processing equipment. Connectors withstand temperatures up to 130°C (266°F) for 60 minutes during SIP.

All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolysulfone
  • Sterilization MethodFull connector assembly: gamma irradiation up to 50 kGy or autoclave up to 129°C (265°F) for 60 minutes
    Steam-in-Place (SIP): up to 130°C (266°F) for 60 minutes
  • Number Of Ports3
  • Connection TypeHose Barb x Sanitary x Sanitary
  • Barb Size / Tubing Id (Port 1) (mm)12.7
  • Barb Size / Tubing Id (Port 1) (in)1/2
  • Sanitary Clamp Size (Port 2) (in)3/4
  • Sanitary Clamp Size (Port 3) (InchShort)3/4
  • Flow PatternStraight
  • ColorAmber tint
  • Min Temperature (° F)39
  • Max Temperature (° F)104
  • Min Temperature (° C)4
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)30
  • Fitting Length (in)5 3/32
  • Fitting Width (in)2
  • Fitting Length (mm)129.3
  • Fitting Width (mm)50.8
  • Regulatory ApprovalsRoHS;Non-Phthalate;ADCF;FDA;USP;Non-DEHP
  • DescriptionSanitary Steam-in-Place Connector, Single Cycle, Polysulfone with Polyethylene Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Ea
Documents & Certificates
Reviews
Q&A