CPC® (Colder) Steam-Thru® II Sanitary Steam-in-Place Connectors, Two Cycle, Polysulfone
Make a quick and easy sterile connection between stainless steel processing equipment and tubing assemblies
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Innovative three-port steam design
  • Offers both “steam on” and “steam off” functionalities
  • Steam-Thru II thumb latch secures valve position
  • 3/4" sanitary connection for condensate port
  • 0.5" nominal flow path size
The CPC® (Colder) Steam-Thru® II connectors feature a unique three-port design for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. Use the Steam-Thru II SIP connection to sterilize the fitting while connected to stainless steel equipment. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle following media transfer. The “steam off” function helps minimize cross-contamination risks associated with reusable components. Select from 3/4" and 1-1/2" sanitary connections for attachment to the processing equipment. Connectors withstand temperatures up to 135°C (275°F) for 60 minutes during SIP.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.
39,300.00 - ₹40,700.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Sanitary Clamp Size (Port 2) (in) Sanitary Clamp Size (Port 3) (InchShort)
Availability Pricing
CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 3/4" Sanitary; 1/Each 5011349 Mfr # STC2020000
COMPARE
1/2 3/4 3/4
₹39,300.00INR / Each
CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 3/8" ID x 3/4" x 3/4" Sanitary; 1/Each 5011350 Mfr # STC2020100
COMPARE
3/8 3/4 3/4
₹39,400.00INR / Each
CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011351 Mfr # STC2020200
COMPARE
1/2 3/4 1-1/2
₹39,500.00INR / Each
CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 3/8" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011352 Mfr # STC2020300
COMPARE
3/8 3/4 1-1/2
₹39,800.00INR / Each
CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 3/4" ID x 3/4" x 3/4" Sanitary; 1/Each 5011353 Mfr # STC2020900
COMPARE
3/4 3/4 3/4
₹40,700.00INR / Each
CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 3/4" ID x 3/4" x 1-1/2" Sanitary; 1/Each 5011354 Mfr # STC2021000
COMPARE
3/4 3/4 1-1/2
₹40,300.00INR / Each