CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 1-1/2" Sanitary; 1/Each

Representative image only.

CPC (Colder) Steam-Thru® II Sanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 1-1/2" Sanitary; 1/Each

CPC (Colder) | Mfr: STC2020200| Item: 5011351

Make a quick and easy sterile connection between stainless steel processing equipment and tubing assemblies

₹39,500.00INR / Each

₹39,500.00INR / Each
Product Description

Key Features

  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Manufactured in an ISO Class 7 certified cleanroom to minimize contamination by pollutants
  • Innovative three-port steam design
  • Offers both “steam on” and “steam off” functionalities
  • Steam-Thru II thumb latch secures valve position
  • 3/4" sanitary connection for condensate port
  • 0.5" nominal flow path size
  • 1/2" hose barb x 1-1/2" sanitary connection

More About this Item

The CPC® (Colder) Steam-Thru® II connectors feature a unique three-port design for a true steam-through steam-in-place (SIP) process, eliminating dead legs and the need for a laminar flow hood. Use the Steam-Thru II SIP connection to sterilize the fitting while connected to stainless steel equipment. The Steam-Thru II connection offers the flexibility of “steam on” and “steam off” functionality, allowing a second SIP cycle following media transfer. The “steam off” function helps minimize cross-contamination risks associated with reusable components. Select from 3/4" and 1-1/2" sanitary connections for attachment to the processing equipment. Connectors withstand temperatures up to 135°C (275°F) for 60 minutes during SIP.

All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications. Fittings are manufactured and sealed in an ISO Class 7 cleanroom to ensure that the integrity is not compromised. An ISO Class 7 cleanroom utilizes HEPA filtration systems to maintain air cleanliness levels of less than 10,000 particles of 0.5 µm or larger per cubic foot.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolysulfone
  • Sterilization MethodFull connector assembly: gamma irradiation up to 50 kGy or autoclave up to 129°C (265°F) for 60 minutes
    Steam-in-Place (SIP): up to 130°C (266°F) for 60 minutes
  • Connection TypeHose Barb x Sanitary x Sanitary
  • Number Of Ports3
  • Barb Size / Tubing Id (Port 1) (mm)12.7
  • Barb Size / Tubing Id (Port 1) (in)1/2
  • Sanitary Clamp Size (Port 2) (in)3/4
  • Sanitary Clamp Size (Port 3) (InchShort)1-1/2
  • Flow PatternStraight
  • ColorAmber tint
  • Min Temperature (° F)39
  • Max Temperature (° F)104
  • Min Temperature (° C)4
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)35
  • Fitting Length (in)6 27/32
  • Fitting Width (in)2 13/32
  • Fitting Length (mm)173.7
  • Fitting Width (mm)61
  • Regulatory ApprovalsNon-Phthalate;ADCF;FDA;USP;RoHS;Non-DEHP
  • DescriptionSanitary Steam-in-Place Connector, Two Cycle, Polysulfone with Polycarbonate Sleeve, 1/2" ID x 3/4" x 1-1/2" Sanitary; 1/Ea
Documents & Certificates
Reviews
Q&A