CPC® (Colder) SMC Mini Quick-Disconnect Fitting, Coupling
Provides more secure alternative to luer-type fittings
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Free coupling rotation eliminates kinked tubing
  • Ideal for tubing sizes from 1/16" to 1/8" ID
  • Acetal or Polypropylene
The CPC (Colder) SMC Series quick-disconnect hose barb couplings offer a twist-to-connect design, providing a reliable and more secure alternative to luer-type fittings. This design allows tubing to rotate freely when connected, preventing both kinked tubing and accidental disconnection during use. The fittings can withstand pressures from vacuum to 100 psi (6.9 bar).
1,700.00 - ₹2,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Connection Type
Availability Pricing
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Coupling, Acetal, Non-Valved, 1/16" ID; 1/Ea 4852738 Mfr # SMC01
COMPARE
1/16 Acetal Quick-Disconnect Hose Barb Coupling, Non-Valved
₹1,800.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Coupling, PP, Non-Valved, 1/16" ID; 1/Ea 4852739 Mfr # SMC0112
COMPARE
1/16 Polypropylene Quick-Disconnect Hose Barb Coupling, Non-Valved
₹2,300.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Coupling, Acetal, Non-Valved, 1/8" ID; 1/Ea 4852740 Mfr # SMC02
COMPARE
1/8 Acetal Quick-Disconnect Hose Barb Coupling, Non-Valved
₹1,700.00INR / Each
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Coupling, PP, Non-Valved, 1/8" ID; 1/Ea 4852741 Mfr # SMC0212
COMPARE
1/8 Polypropylene Quick-Disconnect Hose Barb Coupling, Non-Valved
₹2,100.00INR / Each