CPC (Colder) SMC Mini Quick-Disconnect Fitting, Coupling, PP, Non-Valved, 1/16" ID; 1/Ea
Representative image only.
CPC (Colder) SMC Mini Quick-Disconnect Fitting, Coupling, PP, Non-Valved, 1/16" ID; 1/Ea
CPC (Colder) | Mfr: SMC0112| Item: 4852739
Provides more secure alternative to luer-type fittings
₹2,300.00INR / Each
₹2,300.00INR / Each
Product Description
Key Features
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Free coupling rotation eliminates kinked tubing
  • Ideal for tubing sizes from 1/16" to 1/8" ID
  • Acetal or Polypropylene
  • 1/16" hose barb
More About this Item
The CPC (Colder) SMC Series quick-disconnect hose barb couplings offer a twist-to-connect design, providing a reliable and more secure alternative to luer-type fittings. This design allows tubing to rotate freely when connected, preventing both kinked tubing and accidental disconnection during use. The fittings can withstand pressures from vacuum to 100 psi (6.9 bar).
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Locking Sleeve MaterialAcetal
  • O-Ring MaterialBuna-N
  • Number Of Ports2
  • Connection TypeQuick-Disconnect Hose Barb Coupling, Non-Valved
  • Barb Size / Tubing Id (Port 1) (in)1/16
  • Barb Size / Tubing Id (Port 2) (in)1/16