CPC® (Colder) PMCD Quick-Disconnect Fittings, UNF(F) Body & Insert, Valved, Polypropylene
Resistant to most chemical solutions
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from microbore to 1/4” ID
  • Chemical resistance
  • Gamma Sterilizable
The CPC (Colder) PLCD Series quick disconnect threaded UNF(F) body and insert fittings have a 1/8" flow size, making them suitable for a wide range of general-purpose and chemical applications. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
Constructed of polypropylene, the panel mount fittings with a ¼”-28 flat bottom port, provide enhanced chemical resistance, making them ideal for harsher environments.
An ergonomic thumb latch enables effortless one-handed connection and disconnection, even when wearing gloves. These fittings can withstand pressures ranging from vacuum to 120 psi (8.3 bar) and operate effectively within a temperature range of 32 to 160 °F (0 to 71 °C).
2,900.00 - ₹62,700.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread size UNF(F) (port 1) (InchShort)
Availability Pricing
CPC (Colder) PMCD Quick-Disconnect Fittings, Thread Body, Panel Mount, Valved, Polypropylene, 1/4-28 UNF(F); 25/Pk 4901468 Mfr # PMCD18042812/25 PK
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 0.25
₹62,500.00INR / Pkg of 25
CPC (Colder) PMCD12 Quick-Disconnect Fittings, Thread Insert, Panel Mount, Valved, Polypropylene, 1/4-28 UNF(F); 25/Pk 4901469 Mfr # PMCD48042812/25 PK
COMPARE
Straight Quick-Disconnect Threaded Insert, Panel Mount Valve 0.25
₹62,700.00INR / Pkg of 25
CPC (Colder) PMCD Quick-Disconnect Fittings, Thread Body, Panel Mount, Valved, Polypropylene, 1/4-28 UNF(F) 5013658 Mfr # PMCD18042812
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 0.25
₹2,900.00INR / Each
CPC (Colder) PMCD12 Quick-Disconnect Fittings, Thread Insert, Panel Mount, Valved, Polypropylene, 1/4-28 UNF(F) 5013671 Mfr # PMCD48042812
COMPARE
Straight Quick-Disconnect Threaded Insert, Panel Mount Valve 0.25
₹3,000.00INR / Each