CPC® (Colder) MPC™/MPX® Quick-Disconnect Fittings, Sanitary Insert, Non-Valved, Polysulfone
Connect to a coupling body to quickly create a sterile fluid pathway
  • Connect to a female coupling body for easy media transfer
  • Mix and match termination sizes for added flexibility
  • 0.375" nominal flow path size
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
The CPC® (Colder) MPC™/MPX® series connectors provide a simple way to transfer media between single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. Simply use a cap to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
3,700.00 - ₹4,800.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
CPC (Colder) MPC™ Quick-Disconnect Fitting, Sanitary Insert, Polysulfone, Non-Valved, 3/4" Sanitary; 1/Each 5011283 Mfr # MPC44012T39M
COMPARE
Polysulfone Quick Disconnect MPC Body x Sanitary 3/4
₹3,700.00INR / Each
CPC (Colder) MPC™ Quick-Disconnect Fitting, Sanitary Insert, Polysulfone, Non-Valved, 1-1/2" Sanitary; 1/Each 5011284 Mfr # MPC44024T39M
COMPARE
Polysulfone Quick Disconnect MPC Body x Sanitary 1-1/2
₹4,100.00INR / Each
CPC (Colder) MPX® Quick-Disconnect Fitting, Sanitary Insert, Polysulfone, Non-Valved, 3/4" Sanitary; 1/Each 5011331 Mfr # MPX44012T39M
COMPARE
Polysulfone Quick Disconnect MPX Body x Sanitary 3/4
₹4,100.00INR / Each
CPC (Colder) MPX® Quick-Disconnect Fitting, Sanitary Insert, Polysulfone, Non-Valved, 1-1/2" Sanitary; 1/Each 5011332 Mfr # MPX44024T39M
COMPARE
Polysulfone Quick Disconnect MPX Body x Sanitary 1-1/2
₹4,800.00INR / Each