CPC® (Colder) MPC/MPX Quick-Disconnect Fittings, Back-to-Back Body Adapter
Connect single-use systems that feature identical coupling connections at the end of their tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a male coupling insert for easy media transfer
  • Operate ergonomic thumb latch even with gloved hands
  • Combine MPC and MPX couplings for reducing options
The CPC® (Colder) MPC™/MPX® series back-to-back adapters provide an easy way to convert to industry standard connections or single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. These adapters provide an easy transition between multiple tubing sizes ranging from 1/8" to 1/2" ID. Simply use a plug to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
2,200.00 - ₹5,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Connection Type
Availability Pricing
CPC (Colder) MPC™ Quick-Disconnect Fitting, MPC™ to MPC Back-to-Back Body Adapter, Polycarbonate; 1/Each 5011249 Mfr # MPC17C1703
COMPARE
Purple tint Polycarbonate MPC Body x MPC Body
₹2,200.00INR / Each
CPC (Colder) MPC™ Quick-Disconnect Fitting, MPC™ to MPC Back-to-Back Body Adapter, Polysulfone; 1/Each 5011250 Mfr # MPC17C1739
COMPARE
Amber tint Polysulfone MPC Body x MPC Body
₹2,600.00INR / Each
CPC (Colder) MPC™ Quick-Disconnect Fitting, MPC™ to MPX® Back-to-Back Body Adapter, Polycarbonate; 1/Each 5011251 Mfr # MPC17X1703
COMPARE
Purple tint Polycarbonate MPC Body x MPX Body
₹2,800.00INR / Each
CPC (Colder) MPC™ Quick-Disconnect Fitting, MPC™ to MPX® Back-to-Back Body Adapter, Polysulfone; 1/Each 5011252 Mfr # MPC17X1739
COMPARE
Amber tint Polysulfone MPC Body x MPX Body
₹4,000.00INR / Each
CPC (Colder) MPX® Quick-Disconnect Fitting, MPX® to MPX Back-to-Back Body Adapter, Polycarbonate; 1/Each 5011311 Mfr # MPX17X1703
COMPARE
Purple tint Polycarbonate MPX Body x MPX Body
₹2,800.00INR / Each
CPC (Colder) MPX® Quick-Disconnect Fitting, MPX® to MPX Back-to-Back Body Adapter, Polysulfone; 1/Each 5011312 Mfr # MPX17X1739
COMPARE
Amber tint Polysulfone MPX Body x MPX Body
₹5,400.00INR / Each