CPC® (Colder) MicroCNX® Series Nano Quick-Disconnect Fittings, Genderless Hose Barb
Micro-flow nano connectors simplify sterile tubing connections for small-format bioprocessing
  • Withstand ultra-low temperatures down to -190°C
  • Simple “pinch, click, and pull” design—no tools required
  • Enables fast and error-free connections
  • Tailored for cell and gene therapy (CGT)
  • Designed for micro-flow media transfer applications
  • Free technical application support available to determine which connector type is needed for your application
The MicroCNX® genderless nano connectors offer a fast, simple, and secure solution for connecting tubing in small-format biomanufacturing assemblies. Available with 1/8” and 1/16” hose barb terminations, they are compatible with a wide range of tubing materials including silicone, TPE, and PVC. Engineered to withstand ultra-low temperatures down to -190°C, the connectors are ideal for integration with freezing cassettes used in cell and gene therapy (CGT) processing.
Designed for aseptic operation, the connectors maintain a closed flow path—ensuring sterile connections even in non-sterile environments. They eliminate the need for tube welding, helping reduce setup time and overall costs. Operation is intuitive: pinch to remove the protective cover, align and join the two halves, then squeeze until you hear an audible "click." Finally, pull away the membrane strips to complete the sterile connection.
The connectors are purpose-built for secure, sterile fluid transfer in CGT and other biological media applications.
6,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Max Operating Pressure (PSI)
Availability Pricing
CPC (Colder) MicroCNX® Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/16” ID 5013830 Mfr # CNY17101HT
COMPARE
1/16 Polyphenylsulfone 29
₹6,300.00INR / Each
CPC (Colder) MicroCNX® Nano Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polyphenylsulfone, 1/8” ID 5013831 Mfr # CNY17102HT
COMPARE
1/8 Polyphenylsulfone 29
₹6,300.00INR / Each