CPC (Colder) MCD Quick-Disconnect Fittings, Threaded Body & Insert, Valved, Chrome-Plated Brass
Provides quick and secure connection and disconnection of fluid lines
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly, even with gloved hands
  • Withstands higher pressure and temperature
  • Mix and match termination sizes for added flexibility
  • Compatible with PMC acetal and PMC12 polypropylene couplings
The CPC (Colder) MCD series quick-disconnect threaded body and insert fittings feature an automatic shutoff valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. Simply push the thumb latch to disconnect. The thumb latch is designed so only one hand is required to safely connect and disconnect the fittings quickly, even with gloves on.
These chrome-plated brass fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures.
3,000.00 - ₹30,600.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) MCD Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Valved, 1/8" NPT; 10/Pk 1514511 Mfr # MCD1002/10 PK
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 1/8
₹30,600.00INR / Pkg of 10
CPC (Colder) MCD Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Valved, 1/8" NPT 5013580 Mfr # MCD1002
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 1/8
₹3,000.00INR / Each
CPC (Colder) MCD Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Valved, 1/4" NPT 5013581 Mfr # MCD1004
COMPARE
Straight Quick-Disconnect Threaded Body, Valve 1/4
₹3,100.00INR / Each
CPC (Colder) MCD Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Valved, 1/8" NPT 5013586 Mfr # MCD2402
COMPARE
Straight Quick-Disconnect Threaded Insert, Valve 1/8
₹3,000.00INR / Each
CPC (Colder) MCD Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Valved, 1/4" NPT 5013587 Mfr # MCD2404
COMPARE
Straight Quick-Disconnect Threaded Insert, Valve 1/4
₹3,400.00INR / Each