CPC® (Colder) LCD Quick-Disconnect Fittings, Threaded Body & Insert, Valved, Chrome-Plated Brass
Withstands higher pressures and temperatures
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Ergonomic design makes it easy to connect and disconnect fittings quickly
  • Ideal for tubing sizes from ¼” to 3/8” ID
  • Compatible with PLCD acetal and PLCD12 polypropylene couplings
The CPC (Colder) LCD series quick-disconnect threaded body and insert fittings are built to endure the most demanding applications and are ideal for use with higher temperatures or pressures. The fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. The high-efficiency design delivers superior flow capacity with minimal spillage in a compact size. An ergonomic thumb latch allows you to safely connect and disconnect the fittings with the use of one hand, even with gloves on.
The fittings can operate in temperatures from -40 to 180 °F (-40 to 82 °C) and pressures from vacuum to 250 psi (17.3 bar).
3,600.00 - ₹38,400.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) LCD Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Valved, 1/4" NPT(M); 10/PK 4901299 Mfr # LCD10004/10 PK
COMPARE
Straight Quick-Disconnect Threaded Body, Valved 1/4
₹38,400.00INR / Pkg of 10
CPC (Colder) LCD Quick-Disconnect Fitting, Threaded Body, Chrome-Plated Brass, Valved, 1/4" NPT(M) 5013564 Mfr # LCD10004
COMPARE
Straight Quick-Disconnect Threaded Body, Valved 1/4
₹3,600.00INR / Each
CPC (Colder) LCD Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Valved, 1/4" NPT(M) 5013569 Mfr # LCD24004
COMPARE
Straight Quick-Disconnect Threaded Insert, Valved 1/4
₹3,600.00INR / Each
CPC (Colder) LCD Quick-Disconnect Fitting, Threaded Insert, Chrome-Plated Brass, Valved, 3/8" NPT(M) 5013570 Mfr # LCD24006
COMPARE
Straight Quick-Disconnect Threaded Insert, Valved 3/8
₹3,800.00INR / Each