CPC® (Colder) HFCD35 Quick-Discount Fittings, Threaded Insert, Valved, Polysulfone
Withstands demanding applications while providing reliable performance
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and autoclavable
  • Ideal for tubing sizes from 3/8“ to 3/4” ID
  • Compatible with HFCD12 couplings
The CPC (Colder) HFCD35 Series quick-disconnect threaded insert fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. The high-efficiency design delivers superior flow capacity with minimal spillage in a compact size. Featuring an ergonomic plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation.
Made from chemically resistant polysulfone, they are ideal for harsh environments, providing durability and reliable performance. The fittings can operate in temperatures from -40 to 280 °F (-40 to 138 °C) and pressures from vacuum to 125 psi (8.6 bar).
2,900.00 - ₹3,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Threaded Insert, Polysulfone, Valved, 3/8" NPT(M); 1/Each 5013552 Mfr # HFCD24635
COMPARE
Straight Quick-Disconnect Threaded Insert, Valved 3/8
₹2,900.00INR / Each
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Threaded Insert, Polysulfone, Valved, 1/2" NPT(M); 1/Each 5013554 Mfr # HFCD24835
COMPARE
Straight Quick-Disconnect Threaded Insert, Valved 1/2
₹3,000.00INR / Each