CPC® (Colder) HFCD35 Quick-Discount Fittings, Hose Barb Body & Insert, Valved, Polysulfone
Withstands demanding applications while providing reliable performance
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and autoclavable
  • Ideal for tubing sizes from 3/8“ to 3/4” ID
  • Compatible with HFCD12 couplings
The CPC (Colder) HFCD35 Series quick-disconnect hose barb and insert fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. The high-efficiency design delivers superior flow capacity with minimal spillage in a compact size. Featuring an ergonomic plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation.
Made from chemically resistant polysulfone, they are ideal for harsh environments, providing durability and reliable performance. The fittings can operate in temperatures from -40 to 280 °F (-40 to 138 °C) and pressures from vacuum to 125 psi (8.6 bar).
2,600.00 - ₹3,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 3/8" ID; 1/Each 5013538 Mfr # HFCD17635
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Valved
₹3,800.00INR / Each
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Hose Barb Body, Polysulfone, Valved, 1/2" ID; 1/Each 5013540 Mfr # HFCD17835
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Body, Valved
₹3,800.00INR / Each
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, 3/8" ID; 1/Each 5013544 Mfr # HFCD22635
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Insert, Valved
₹2,600.00INR / Each
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Valved, 1/2" ID; 1/Each 5013546 Mfr # HFCD22835
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Insert, Valved
₹2,600.00INR / Each
CPC (Colder) HFCD35 Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Polysulfone, Valved, 1/2" ID; 1/Each 5013549 Mfr # HFCD23835
COMPARE
1/2 Elbow Quick-Disconnect Hose Barb Insert, Valved
₹3,300.00INR / Each