CPC® (Colder) HFCD12 Quick-Disconnect Fittings, Hose Barb Body & Insert, Valved, Polypropylene
High efficiency design provides more flow and lower spillage
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 3/8“ to 3/4” ID
  • Compatible with HFCD35 couplings
The CPC (Colder) HFCD12 Series quick-disconnect hose barb body and insert fittings feature an automatic shutoff spring-operated stainless-steel valve which closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. The high-efficiency design delivers superior flow capacity with minimal spillage in a compact size. Featuring an ergonomic plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation.
Made from chemically resistant polypropylene, they are ideal for harsh environments, providing durability and reliable performance. The fittings can operate in temperatures from 32 to 160 °F (0 to 71 °C) and pressures from vacuum to 60 psi (4.2 bar).
2,300.00 - ₹42,300.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 3/8" ID; 10/Pk 3121301 Mfr # HFCD17612/10 PK
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Valve
₹33,300.00INR / Pkg of 10
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Polypropylene, Valved, 1/2" ID; 10/Pk 4901288 Mfr # HFCD16812/10 PK
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Body, Panel Mount, Valve
₹42,300.00INR / Pkg of 10
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Panel Mount, Polypropylene, Valved, 1/2" ID 5013534 Mfr # HFCD16812
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Body, Panel Mount, Valve
₹3,900.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 5/8" ID 5013535 Mfr # HFCD171012
COMPARE
5/8 Straight Quick-Disconnect Hose Barb Body, Valve
₹3,400.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 3/4" ID 5013536 Mfr # HFCD171212
COMPARE
3/4 Straight Quick-Disconnect Hose Barb Body, Valve
₹3,100.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 3/8" ID 5013537 Mfr # HFCD17612
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Valve
₹3,000.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 1/2" ID 5013539 Mfr # HFCD17812
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Body, Valve
₹3,100.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 5/8" ID 5013541 Mfr # HFCD221012
COMPARE
5/8 Straight Quick-Disconnect Hose Barb Insert, Valve
₹2,500.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 3/4" ID 5013542 Mfr # HFCD221212
COMPARE
3/4 Straight Quick-Disconnect Hose Barb Insert, Valve
₹2,400.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 3/8" ID 5013543 Mfr # HFCD22612
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Insert, Valve
₹2,300.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Valved, 1/2" ID 5013545 Mfr # HFCD22812
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Insert, Valve
₹2,300.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Polypropylene, Valved, 3/8" ID 5013547 Mfr # HFCD23612
COMPARE
3/8 Elbow Quick-Disconnect Hose Barb Insert, Valve
₹2,800.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Polypropylene, Valved, 1/2" ID 5013548 Mfr # HFCD23812
COMPARE
1/2 Elbow Quick-Disconnect Hose Barb Insert, Valve
₹2,800.00INR / Each
CPC (Colder) HFCD12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Valved, 3/4" ID; 10/Pk 6123309 Mfr # HFCD171212/10 PK
COMPARE
3/4 Straight Quick-Disconnect Hose Barb Body, Valve
₹37,300.00INR / Pkg of 10
Showing 1- 14 of 14