CPC® (Colder) HFC39 Quick-Disconnect Fittings, Hose Barb Insert, Polysulfone
Make sterile connections quick and easy with the push-to-connect fittings
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a female coupling body for easy fluid transfer
  • Audible “click” provides added assurance of a secure connection
  • Quick one-step disconnection process minimize user error
Connect a male insert to a female body for a secure fluid flow. These CPC® (Colder) HFC39 quick-disconnect hose barb inserts are available with or without an automatic shutoff valve. An automatic shutoff valve closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. Simply push the thumb latch to disconnect.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
2,900.00 - ₹4,100.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 1/4" ID; 1/Each 5011217 Mfr # HFC22439M
COMPARE
1/4 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 1/4" ID; 1/Ea
₹2,900.00INR / Each
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 3/8" ID; 1/Each 5011218 Mfr # HFC22639M
COMPARE
3/8 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 3/8" ID; 1/Ea
₹2,900.00INR / Each
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 1/2" ID; 1/Each 5011219 Mfr # HFC22839M
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone, Non-Valved, 1/2" ID; 1/Ea
₹2,900.00INR / Each
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Stainless Steel Valve, 1/4" ID; 1/Each 5011228 Mfr # HFCD22439M
COMPARE
1/4 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Stainless Steel Valve, 1/4" ID; 1/Ea
₹4,100.00INR / Each
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Stainless Steel Valve, 1/2" ID; 1/Each 5011232 Mfr # HFCD22839M
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Stainless Steel Valve, 1/2" ID; 1/Ea
₹4,100.00INR / Each