CPC® (Colder) HFC12 Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved, Polypropylene
High efficiency design provides more flow and lower spillage
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Chemically resistant and gamma sterilizable
  • Ideal for tubing sizes from 3/8“ to 3/4” ID
  • Compatible with HFC35 couplings
The CPC (Colder) HFC12 Series quick-disconnect hose barb body and insert fittings offer a high-efficiency design, delivering superior flow capacity with minimal spillage in a compact size. Featuring an ergonomic plastic thumb latch with no exposed metal, these fittings ensure easy, gloved-hand operation.
Made from chemically resistant polypropylene, they are ideal for harsh environments, providing durability and reliable performance. The fittings can operate in temperatures from 32 to 160 °F (0 to 71 °C) and pressures from vacuum to 60 psi (4.2 bar).
1,100.00 - ₹2,600.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) HFC12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Non-Valved, 3/8" ID; 1/Each 5013522 Mfr # HFC17612
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Non-Valved
₹2,400.00INR / Each
CPC (Colder) HFC12 Quick-Disconnect Fitting, Hose Barb Body, Polypropylene, Non-Valved, 1/2" ID; 1/Each 5013523 Mfr # HFC17812
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹2,600.00INR / Each
CPC (Colder) HFC12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Non-Valved, 5/8" ID; 1/Each 5013524 Mfr # HFC221012
COMPARE
5/8 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹1,200.00INR / Each
CPC (Colder) HFC12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Non-Valved, 3/8" ID; 1/Each 5013525 Mfr # HFC22612
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹1,100.00INR / Each
CPC (Colder) HFC12 Quick-Disconnect Fitting, Hose Barb Insert, Polypropylene, Non-Valved, 1/2" ID; 1/Each 5013527 Mfr # HFC22812
COMPARE
1/2 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹1,100.00INR / Each
CPC (Colder) HFC12 Quick-Disconnect Fitting, Hose Barb Insert, Elbow, Polypropylene, Non-Valved, 1/2" ID; 1/Each 5013529 Mfr # HFC23812
COMPARE
1/2 Elbow Quick-Disconnect Hose Barb Insert, Non-Valved
₹1,800.00INR / Each