CPC® (Colder) HFC Quick-Disconnect Fittings, Hose Barb Insert, Polysulfone with Alloy C-276 Valve
Valved connectors ensure sterility is maintained on both sides of the system during the disconnection process
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a female coupling body for easy fluid transfer
  • Audible “click” provides added assurance of a secure connection
  • Quick one-step disconnection process minimize user error
  • Alloy C-276 valve spring stops the flow and offers broad application compatibility
  • Laser-etched item and lot numbers offer batch traceability
The CPC® (Colder) HFC series connectors feature an automatic shutoff valve enabling quick disconnection while maintaining its sterility. Connect a male insert and a female body for a secure fluid flow. Simply push the thumb latch to disconnect. An alloy C-276 valve spring in each connector closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system. Laser-etched item and lot numbers provide batch traceability.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
5,900.00 - ₹6,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) HFC Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Alloy C-276 Valve, 1/4" ID; 1/Each 5011229 Mfr # HFCD22439MHC
COMPARE
1/4 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Alloy C-276 Valve, 1/4" ID; 1/Ea
₹6,000.00INR / Each
CPC (Colder) HFC Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Alloy C-276 Valve, 3/8" ID; 1/Each 5011231 Mfr # HFCD22639MHC
COMPARE
3/8 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Alloy C-276 Valve, 3/8" ID; 1/Ea
₹5,900.00INR / Each
CPC (Colder) HFC Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Alloy C-276 Valve, 1/2" ID; 1/Each 5011233 Mfr # HFCD22839MHC
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting, Hose Barb Insert, Polysulfone with Alloy C-276 Valve, 1/2" ID; 1/Ea
₹6,000.00INR / Each