CPC® (Colder) APC Quick-Disconnect Fittings, Threaded Body, Non-Valved, Acetal
Connect to a quick-disconnect fitting insert to swiftly create a secure fluid pathway
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Shrouded thumb latch protects against accidental disconnects
  • Ideal for tubing sizes from 1/4“ to 3/8” ID
  • Compatible with most EFC couplings
The CPC (Colder) APC series quick-disconnect threaded body fittings are designed for ease of use and efficiency. The plastic thumb latch features no exposed metal parts and is easy to operate even with gloved hands. The fittings' fewer moving parts ensure reliability and reduce the risk of wear or failure over time. Their contoured design not only enhances usability but also promotes excellent flow characteristics.
With integral terminations, the fittings minimize leak points, eliminate the need for extra components, and enable faster installations with shorter assemblies. Ideal for single-use systems, they offer a simple, reliable way to transfer media with minimal complexity.
690.00 - ₹760.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Connection Type Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
CPC (Colder) APC Quick-Disconnect Fitting, Threaded Body, Acetal, Non-Valved, 1/4" NPT(M); 1/Each 5013510 Mfr # APC10004
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valve 1/4
₹690.00INR / Each
CPC (Colder) APC Quick-Disconnect Fitting, Threaded Body, Acetal , Non-Valved, 3/8" NPT(M); 1/Each 5013511 Mfr # APC10006
COMPARE
Straight Quick-Disconnect Threaded Body, Non-Valve 3/8
₹760.00INR / Each