CPC® (Colder) APC Quick-Disconnect Fittings, Hose Barb Body & Insert, Non-Valved, Acetal
Provides quick and secure connection and disconnection of fluid lines
  • Ideal for general purposes, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support is available to determine which connector type is needed for your application
  • Shrouded thumb latch protects against accidental disconnects
  • Ideal for tubing sizes from 1/4“ to 3/8” ID
  • Compatible with most EFC couplings
The CPC (Colder) APC series quick-disconnect hose barb body and insert fittings are designed for ease of use and efficiency. The plastic thumb latch features no exposed metal parts and is easy to operate even with gloved hands. The fittings' fewer moving parts ensure reliability and reduce the risk of wear or failure over time. Their contoured design not only enhances usability but also promotes excellent flow characteristics.
With integral terminations, the fittings minimize leak points, eliminate the need for extra components, and enable faster installations with shorter assemblies. Ideal for single-use systems, they offer a simple, reliable way to transfer media with minimal complexity.
230.00 - ₹104,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Flow Pattern Connection Type
Availability Pricing
CPC (Colder) APC Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 1/4" ID; 250/PK 4901555 Mfr # APC17004 250/PK
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹87,500.00INR / Pkg of 250
CPC (Colder) APC Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 1/4" ID; 1/Each 5013512 Mfr # APC17004
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹390.00INR / Each
CPC (Colder) APC Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 3/8" ID; 1/Each 5013513 Mfr # APC17006
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹410.00INR / Each
CPC (Colder) APC Quick-Disconnect Fitting, Hose Barb Insert, Acetal, Non-Valved, 1/4" ID; 1/Each 5013514 Mfr # APC22004
COMPARE
1/4 Straight Quick-Disconnect Hose Barb Insert, Non-Valve
₹230.00INR / Each
CPC (Colder) APC Quick-Disconnect Fitting, Hose Barb Body, Acetal, Non-Valved, 3/8" ID; 250/CS 6123191 Mfr # APC17006/250 CS
COMPARE
3/8 Straight Quick-Disconnect Hose Barb Body, Non-Valve
₹104,000.00INR / Case of 250