Corrosive Substance Storage Cabinets
Reduce fire risks and protect workers
  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
Constructed of chemical-resistant, 8-mm polypropylene (PP) panels offering excellent protection against harsh acids, alkalis, and corrosion. Cabinets are divided into two parts for different
substances, making good use of storage space. (Not shown)
Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and another on the bottom effectively reduce the concentration of volatile substances and prevent fire hazards.
Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.
44,900.00 - ₹191,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Number of Shelves Description
Availability Pricing
SYSBEL ACP810004 4 Gal 1597697 Mfr # ACP810004
COMPARE
15 1 Corrosive Substance Storage Cabinets; 4 Gal
₹44,900.00INR / Each
SYSBEL ACP810030 30 Gal 1597698 Mfr # ACP810030
COMPARE
114 1 Corrosive Substance Storage Cabinets; 30 Gal
₹120,000.00INR / Each
SYSBEL ACP810012 12 Gal 1597699 Mfr # ACP810012
COMPARE
45 1 Corrosive Substance Storage Cabinets; 12 Gal
₹71,000.00INR / Each
SYSBEL ACP810048 48 Gal 1597700 Mfr # ACP810048
COMPARE
182 3 Corrosive Substance Storage Cabinets; 48 Gal
₹163,000.00INR / Each
SYSBEL ACP810048T 48 Gal 1598526 Mfr # ACP810048T
COMPARE
182 3 Corrosive Substance Storage Cabinets; 48 Gal
₹191,000.00INR / Each