SYSBEL ACP810048T 48 Gal

Representative image only.

SYSBEL ACP810048T 48 Gal

SYSBEL | Mfr: ACP810048T| Item: 1598526

Reduce fire risks and protect workers

₹191,000.00INR / Each

₹191,000.00INR / Each
Product Description

Includes

Shelf

Key Features

  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
  • 15 month warranty

More About this Item

Constructed of chemical-resistant, 8-mm polypropylene (PP) panels offering excellent protection against harsh acids, alkalis, and corrosion. Cabinets are divided into two parts for different
substances, making good use of storage space. (Not shown)

Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and another on the bottom effectively reduce the concentration of volatile substances and prevent fire hazards.
Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.

15 Month Warranty
Specifications
  • Material8mmPP Polypropylene
  • Capacity (Liters)182
  • Capacity (Gal)48
  • Number of Shelves3
  • Width (in)42 29/32
  • Height (in)70 29/32
  • Depth (InchShort)18 1/10
  • Width (cm)109
  • Height (cm)180
  • Depth (cm)46
  • ColorWhite
  • ComplianceCE Cerification
  • Shipping Weight (lbs)133
  • DescriptionCorrosive Substance Storage Cabinets; 48 Gal
  • Warranty15 month
Reviews
Q&A