Corrosive Cabinets

Reduce fire risks and protect workers
  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
Use these cabinets for storing corrosive chemicals which are also
flammable (low and median flash point liquid, flash point <37.8).

Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier
construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and
another on the bottom) effectively reduce the concentration of volatile substances and prevent fire hazards.
Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The
manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be
padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if
a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.
46,400.00 - ₹148,000.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Number of Shelves Description
Availability Pricing
SYSBEL WA810040B 4 Gal/15L,1 door, manual 1597685 Mfr # WA810040B
COMPARE
15 1 Corrosive Cabinets; 4 Gal/15L,1 door, manual
₹46,400.00INR / Each
SYSBEL WA810120B 12 Gal/45L,1 door, manual 1597686 Mfr # WA810120B
COMPARE
45 1 Corrosive Cabinets; 12 Gal/45L,1 door, manual
₹80,900.00INR / Each
SYSBEL WA810300B 30 Gal/114L,2 door, manual 1597687 Mfr # WA810300B
COMPARE
114 1 Corrosive Cabinets; 30 Gal/114L,2 door, manual
₹97,700.00INR / Each
SYSBEL WA810450B 45 Gal/170L,2 door, manual 1597688 Mfr # WA810450B
COMPARE
170 2 Corrosive Cabinets; 45 Gal/170L,2 door, manual
₹123,000.00INR / Each
SYSBEL WA810860B 90 Gal/340L,2 door, manual 1597689 Mfr # WA810860B
COMPARE
340 2 Corrosive Cabinets; 90 Gal/340L,2 door, manual
₹148,000.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat