SYSBEL WA810450B 45 Gal/170L,2 door, manual

Representative image only.

SYSBEL WA810450B 45 Gal/170L,2 door, manual

SYSBEL | Mfr: WA810450B| Item: 1597688

Reduce fire risks and protect workers

₹123,000.00INR / Each

₹123,000.00INR / Each
Product Description

Includes

Shelf

Key Features

  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
  • 10 year warranty

More About this Item

Use these cabinets for storing corrosive chemicals which are also
flammable (low and median flash point liquid, flash point <37.8).

Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier
construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and
another on the bottom) effectively reduce the concentration of volatile substances and prevent fire hazards.
Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The
manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be
padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if
a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.

10 Year Warranty
Specifications
  • MaterialSteel
  • Capacity (Liters)170
  • Capacity (Gal)45
  • Number of Shelves2
  • Width (in)42 29/32
  • Height (in)65
  • Depth (InchShort)18 1/10
  • Width (cm)109
  • Height (cm)165
  • Depth (cm)46
  • ColorBlue
  • ComplianceFM, CE Cerification
  • Shipping Weight (lbs)133
  • DescriptionCorrosive Cabinets; 45 Gal/170L,2 door, manual
  • Warranty10 year
Reviews
Q&A