Corning Multiposition Stirrers 15299

  • High speed—up to 2000 rpm
  • Stir up to nine vessels at the same time
  • Remote control models available—ideal for use in incubators
Multiposition glass-filled resin stirrers available in four, five, and nine position models. Stirrer magnets and reliable DC motor provide smooth, well coupled stirringup to 2000 rpm. The speed of all positions on units are set simultaneously with either the integral knob or remote controller. Remote control ideal for use in incubators.
436,000.00 - ₹584,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) No. Of Stirring Positions Power (VAC)
Availability Pricing
Corning 440824 Four-Position Stirrer, 120 VAC, 60 Hz 8430400 Mfr # 440824
COMPARE
2000 4 120
₹436,000.00INR / Each
Corning 440825 Five-Position Stirrer, 120 VAC, 60 Hz 8430410 Mfr # 440825
COMPARE
2000 5 120
₹470,000.00INR / Each
Corning 440826 Nine-Position Stirrer, 120 VAC, 60 Hz 8430420 Mfr # 440826
COMPARE
2000 9 120
₹584,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat