Corning Internal Thread Cryogenic Vials, with Washer Seals, 500/case
Silicone Washer for Highest level of leak proof assurance and security
  • Supplied Sterile and Certified Non-Pyrogenic
  • Polypropylene construction withstands temperatures to -196°C.
  • Easier to read black graduations and larger marking spot
Vials accept Corning® color-coded cap inserts. Self-standing models lock into Corning® cryogenic racks for single-handed operation. Order colored cap inserts separately.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Inside Diameter (ID) (in) Outside Diameter (OD) (in) Volume (mL)
Availability Pricing
Corning 430487 Internal Thread Cryogenic Vials; 1.2 mL, Washer Seal, 500/cs WW-15941-75 Mfr # 430487
COMPARE
Call For Price
Corning 430488 Internal Thread Cryogenic Vials; 2.0 mL, Washer Seal, Self-Standing WW-15941-76 Mfr # 430488
COMPARE
Call For Price
Corning 430489 Internal Thread Cryogenic Vials; 2.0 mL, Washer Seal, Not Self-Standing WW-15941-77 Mfr # 430489
COMPARE
Call For Price
Corning 430490 Internal Thread Cryogenic Vials; 4.0 mL, Washer Seal, Not Self-Standing WW-15941-78 Mfr # 430490
COMPARE
Call For Price
Corning 430491 Internal Thread Cryogenic Vials; 4.0 mL, Washer Seal, Self-Standing WW-15941-79 Mfr # 430491
COMPARE
Call For Price
Corning 430492 Internal Thread Cryogenic Vials; 5.0 mL, Washer Seal, Not Self-Standing WW-15941-80 Mfr # 430492
COMPARE
Call For Price
Corning 430656 Internal Thread Cryogenic Vials; 5.0 mL, Washer Seal, Self-Standing WW-15941-81 Mfr # 430656
COMPARE
Call For Price
Showing 1- 7 of 7