Corning 430488 Internal Thread Cryogenic Vials; 2.0 mL, Washer Seal, Self-Standing
Representative image only.
Corning 430488 Internal Thread Cryogenic Vials; 2.0 mL, Washer Seal, Self-Standing
Corning | Mfr: 430488| Item: 1594176
Silicone Washer for Highest level of leak proof assurance and security
Call For Price
Call For Price
Product Description
Key Features
  • Supplied Sterile and Certified Non-Pyrogenic
  • Polypropylene construction withstands temperatures to -196°C.
  • Easier to read black graduations and larger marking spot
More About this Item
Vials accept Corning® color-coded cap inserts. Self-standing models lock into Corning® cryogenic racks for single-handed operation. Order colored cap inserts separately.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
  • Tube Size2.0 mL
  • DescriptionInternal Thread Cryogenic Vials; 2.0 mL, Washer Seal, Self-Standing