Corning® Gosselin™ Octagonal PET Storage Bottles
Lightweight and unbreakable media bottles
  • See-through as glass with molded graduations Ergonomic shape for improved handling Wide neck and tamper-evident leakproof screw cap Sterilized fluid path to avoid cross-contamination
Natural PET storage bottles are perfect for contact with food, a sampling of media, biologicals, and other aqueous solutions. All bottles are sterile and come with a tamper-evident 32-mm HDPE screw cap with inner lip.
Bottles arrive at your site enclosed in a PE bag packed inside a box available in two choices. The first choice is in bulk packaging. The tamper-evident caps come fully screwed on with the ring activated. It is bound with film wrap and one traceability label. The second choice is in cardboard trays packaging; bottles are sorted in trays of 12 or 24 with a retractable film wrap and a traceability label for each tray. The tamper-evident caps come loosely screwed on with the ring not yet activated.
46,000.00 - ₹116,000.00INR / Case of 48
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Special Features
Availability Pricing
Corning Gosselin Octagonal Storage Bottle, sterile PET, graduated, bulk pack, 1000 mL; 48/cs 3455107 Mfr # P1000B-50
COMPARE
1000 Bulk pack
₹49,600.00INR / Case of 48
Corning Gosselin Octagonal Storage Bottle, sterile PET, graduated, bulk pack, 125 mL; 336/cs 3455111 Mfr # P125B-50
COMPARE
125 Bulk pack
₹116,000.00INR / Case of 336
Corning Gosselin Octagonal Storage Bottle, sterile PET, graduated, bulk pack, 250 mL; 144/cs 3455113 Mfr # P250B-50
COMPARE
250 Bulk pack
₹70,600.00INR / Case of 144
Corning Gosselin Octagonal Storage Bottle, sterile PET, graduated, bulk pack, 500 mL; 120/cs 3455114 Mfr # P500B-50
COMPARE
500 Bulk pack
₹46,000.00INR / Case of 120