Corning External Thread Cryogenic Vials 61532
External threaded design reduces risk of sample contamination
  • Supplied Sterile and Certified Non-Pyrogenic
  • Polypropylene construction withstands temperatures to -196°C.
  • Silicone Washer for Highest level of leak proof assurance and security
  • Easier to read black graduations and larger marking spot
Vials accept Corning® color-coded cap inserts. Self-standing models lock into Corning® cryogenic racks for single-handed operation. Order cap inserts separately.
Call For Price
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Inside Diameter (ID) (in) Outside Diameter (OD) (in) Volume (mL)
Availability Pricing
Corning 430658 External Thread Cryogenic Vials; 1.2 mL, 500/cs WW-15941-70 Mfr # 430658
COMPARE
Call For Price
Corning 430661 External Thread Cryogenic Vials; 2.0 mL, Not Self-Standing, 500/cs WW-15941-71 Mfr # 430661
COMPARE
Call For Price
Corning 430659 External Thread Cryogenic Vials; 2.0 mL, Self-Standing, 500/cs WW-15941-72 Mfr # 430659
COMPARE
Call For Price
Corning 430662 External Thread Cryogenic Vials; 4.0 mL, 500/cs WW-15941-73 Mfr # 430662
COMPARE
Call For Price
Corning 430663 External Thread Cryogenic Vials; 5.0 mL, 500/cs WW-15941-74 Mfr # 430663