Corning 430661 External Thread Cryogenic Vials; 2.0 mL, Not Self-Standing, 500/cs
Representative image only.
Corning 430661 External Thread Cryogenic Vials; 2.0 mL, Not Self-Standing, 500/cs
Corning | Mfr: 430661| Item: 1594171
External threaded design reduces risk of sample contamination
Call For Price
Call For Price
Product Description
Key Features
  • Supplied Sterile and Certified Non-Pyrogenic
  • Polypropylene construction withstands temperatures to -196°C.
  • Silicone Washer for Highest level of leak proof assurance and security
  • Easier to read black graduations and larger marking spot
More About this Item
Vials accept Corning® color-coded cap inserts. Self-standing models lock into Corning® cryogenic racks for single-handed operation. Order cap inserts separately.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
BioPyr
Specifications
  • Tube Size2.0 mL
  • Description<reg> External Thread Cryogenic Vials; 2.0 mL, Not Self-Standing, 500/cs