Corning® CoolBox™ XT Ice-Free Coolers with One Cooling Core
No mess, no maintenance
  • Keep samples cold without melting ice contamination, inconvenient batteries, or power cord clutter
  • Cooling core maintains sample temperatures at 0.5°C to 4°C for at least four hours
  • Compatible with all Corning CoolSink® and CoolRack® sample modules (order separately)
These benchtop coolers eliminate possible contamination from ice, maintenance costs associated with battery replacement, and messy cords. Simply freeze the cooling core in a -20°C freezer for 12 or more hours, then remove and wait for core to reach 1°C before placing in holder. Once the core is installed, add your CoolSink or CoolRack module and load your samples. Samples quickly equilibrate to the core temperature and remain chilled for 4 to 16 hours.
Select the CoolBox system for longer-term cooling, which features a deeper sample area capable of maintaining temperature up to 16 hours.
225,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color
Availability Pricing
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, purple, 1/ea 0439221 Mfr # 432021
COMPARE
Purple
₹225,000.00INR / Each
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, green, 1/ea 0439222 Mfr # 432022
COMPARE
Green
₹225,000.00INR / Each
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, orange, 1/ea 0439223 Mfr # 432023
COMPARE
Orange
₹225,000.00INR / Each
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, pink, 1/ea 0439224 Mfr # 432024
COMPARE
Pink
₹225,000.00INR / Each