Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, pink, 1/ea
Representative image only.
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, pink, 1/ea
Corning | Mfr: 432024| Item: 0439224
No mess, no maintenance
₹225,000.00INR / Each
₹225,000.00INR / Each
Buy It Together
Current Item
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, pink, 1/ea
Corning XT CoolBox™ Ice-Free Cooler with One Cooling Core, pink, 1/ea
0439224
₹225,000.00INR / Each
+ =
Total Price 252500.00 ADD 2 ITEMS TO CART
Product Description
Includes
Cooling core and removable lid.
Key Features
  • Keep samples cold without melting ice contamination, inconvenient batteries, or power cord clutter
  • Cooling core maintains sample temperatures at 0.5°C to 4°C for at least four hours
  • Compatible with all Corning CoolSink® and CoolRack® sample modules (order separately)
  • 1 year warranty
More About this Item
These benchtop coolers eliminate possible contamination from ice, maintenance costs associated with battery replacement, and messy cords. Simply freeze the cooling core in a -20°C freezer for 12 or more hours, then remove and wait for core to reach 1°C before placing in holder. Once the core is installed, add your CoolSink or CoolRack module and load your samples. Samples quickly equilibrate to the core temperature and remain chilled for 4 to 16 hours.
Select the CoolBox system for longer-term cooling, which features a deeper sample area capable of maintaining temperature up to 16 hours.
1 Year Warranty
Specifications
  • Lowest Set Temperature (° C)-20
  • ColorPink
  • Width (in)7 7/8
  • Width (cm)20
  • DescriptionModule with One Cooling Core, pink, 1/ea