Conical-Bottom Tank Systems

Tank systems are impact and chemical resistant
  • Withstand continuous operating temperature of 150°>F (65°>C)
  • Translucent sides for easy viewing
Linear polyethelyne tanks have conical bottoms for complete drainage of fluids and are impact and chemical resistant. Tanks can withstand continuous operating temperature of 150°>F (65°>C). Translucent sides let you observe liquid levels.
Complete tank systems include tank, carbon steel stand with agitator mounting, LDPE cover, and bulkhead fitting—indicate fitting placement at time of order. Order replacement tanks separately.
276,000.00 - ₹632,000.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Temperature (° F) Volume (Gal)
Availability Pricing
Chem-Tainer Conical-Bottom Tank System, LDPE, 15 gallon 0692500 Mfr # TC1519CA-15GAL FA0100MO-TC1519CM
COMPARE
Polyethylene 150 15
₹276,000.00INR / Each
Chem-Tainer Conical-Bottom Tank System, LDPE, 30 gallon 0692505 Mfr # TC18X29CA-30GAL FA0100MO-TC1829CM
COMPARE
Polyethylene 150 30
₹392,000.00INR / Each
Chem-Tainer Conical-Bottom Tank System, LDPE, 55 gallon 0692510 Mfr # TC2236CA-55GAL FA0100MO-TC2236AS-TC2236CM
COMPARE
Polyethylene 150 55
₹380,000.00INR / Each
Chem-Tainer Conical-Bottom Tank System, LDPE, 100 gallon 0692515 Mfr # TC3036CA-100GAL FA0150MO-TC3036AF-TC3036CM
COMPARE
Polyethylene 150 100
₹632,000.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat