Compression Union Elbow Fittings, PVDF, 5/pack
For corrosive chemical-feed and water-treatment tubing connections
  • Excellent chemical and abrasion resistance
  • Ideal for chemical-feed, water-treatment, filtration, RO/DI, and sample-line connections
  • Temperature range for fittings: -40 to 121°C (-40 to 250°F)
  • Plastic gripper nut helps secure plastic tubing
  • NSF Listed, FDA compliant, and autoclavable
JACO PVDF bulkhead compression union elbow fittings connect rigid plastic tubing to equipment ports in demanding chemical-feed, water-treatment, filtration, RO/DI, and corrosive-fluid systems. They offer excellent abrasion and chemical resistance for applications exposed to corrosive liquids, oxidizers, acids, hydrocarbons, and selected solvents.
3,700.00 - ₹7,100.00INR / Pkg of 5
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in)
Availability Pricing
COMPARE
1/4
₹3,700.00INR / Pkg of 5
COMPARE
3/8
₹5,600.00INR / Pkg of 5
COMPARE
1/2
₹7,100.00INR / Pkg of 5