Compression Fitting, Elbow Union, PVDF, 1/4" OD; 5/Pk
Representative image only.
Compression Fitting, Elbow Union, PVDF, 1/4" OD; 5/Pk
| Item: 3623440
For corrosive chemical-feed and water-treatment tubing connections
₹3,700.00INR / Pkg of 5
₹3,700.00INR / Pkg of 5
Product Description
Key Features
  • Excellent chemical and abrasion resistance
  • Ideal for chemical-feed, water-treatment, filtration, RO/DI, and sample-line connections
  • Temperature range for fittings: -40 to 121°C (-40 to 250°F)
  • Plastic gripper nut helps secure plastic tubing
  • NSF Listed, FDA compliant, and autoclavable
  • 1/4" OD
More About this Item
JACO PVDF bulkhead compression union elbow fittings connect rigid plastic tubing to equipment ports in demanding chemical-feed, water-treatment, filtration, RO/DI, and corrosive-fluid systems. They offer excellent abrasion and chemical resistance for applications exposed to corrosive liquids, oxidizers, acids, hydrocarbons, and selected solvents.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Connection TypeCompression
  • Number Of Ports2
  • Compression Size / Tube Od (Port 1) (in)1/4
  • Compression Size / Tube Od (Port 2) (in)1/4
  • ColorNatural
  • Min Temperature (° F)-40
  • Max Temperature (° F)250
  • Min Temperature (° C)-40
  • Max Temperature (° C)121
  • Max Operating Pressure (PSI)220
  • Quantity5
  • DescriptionCompression Fitting, Elbow Union, PVDF, 1/4" OD; 5/Pk