Compression to NPT(M) Fittings, Straight Adapter, PVDF; 5/Pk
For corrosive chemical-feed and water-treatment tubing connections
  • Excellent chemical and abrasion resistance
  • Ideal for chemical-feed, water-treatment, filtration, RO/DI, and sample-line connections
  • Temperature range for fittings: -40 to 121°C (-40 to 250°F)
  • Plastic gripper nut helps secure plastic tubing
  • NSF Listed, FDA compliant, and autoclavable
JACO PVDF compression to male thread adapters connect rigid plastic tubing to threaded equipment ports in demanding chemical-feed, water-treatment, filtration, RO/DI, and corrosive-fluid systems. They offer excellent abrasion and chemical resistance for applications exposed to corrosive liquids, oxidizers, acids, hydrocarbons, and selected solvents.
PVDF provides strong durability, aging resistance, and chemical compatibility across a broad working range, making it a dependable choice for pharmaceutical, biotech, and food and beverage process-support applications where PVDF compatibility and required certifications have been verified.
These fittings have a semi-translucent, cloudy off-white appearance. PVDF complies with FDA food-contact requirements under 21 CFR § 177.2510, is NSF Listed, and autoclavable.
5,300.00 - ₹9,600.00INR / Pkg of 5
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/4 1/4
₹5,300.00INR / Pkg of 5
COMPARE
3/8 3/8
₹8,000.00INR / Pkg of 5
COMPARE
1/2 1/2
₹9,600.00INR / Pkg of 5