Compression to Threaded Fitting, Straight Adapter, PVDF, 1/4" OD x 1/4" NPT(M); 5/Pk
Representative image only.
Compression to Threaded Fitting, Straight Adapter, PVDF, 1/4" OD x 1/4" NPT(M); 5/Pk
| Item: 3623437
For corrosive chemical-feed and water-treatment tubing connections
₹5,300.00INR / Pkg of 5
₹5,300.00INR / Pkg of 5
Product Description
Key Features
  • Excellent chemical and abrasion resistance
  • Ideal for chemical-feed, water-treatment, filtration, RO/DI, and sample-line connections
  • Temperature range for fittings: -40 to 121°C (-40 to 250°F)
  • Plastic gripper nut helps secure plastic tubing
  • NSF Listed, FDA compliant, and autoclavable
  • 1/4" Tube OD x 1/4" NPT(M)
More About this Item
JACO PVDF compression to male thread adapters connect rigid plastic tubing to threaded equipment ports in demanding chemical-feed, water-treatment, filtration, RO/DI, and corrosive-fluid systems. They offer excellent abrasion and chemical resistance for applications exposed to corrosive liquids, oxidizers, acids, hydrocarbons, and selected solvents.
PVDF provides strong durability, aging resistance, and chemical compatibility across a broad working range, making it a dependable choice for pharmaceutical, biotech, and food and beverage process-support applications where PVDF compatibility and required certifications have been verified.
These fittings have a semi-translucent, cloudy off-white appearance. PVDF complies with FDA food-contact requirements under 21 CFR § 177.2510, is NSF Listed, and autoclavable.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Connection TypeCompression
  • Number Of Ports2
  • Thread Size NPT(M) (Port 1) (InchShort)1/4
  • Compression Size / Tube Od (Port 1) (in)1/4
  • Flow PatternStraight
  • ColorNatural
  • Min Temperature (° F)-40
  • Max Temperature (° F)250
  • Min Temperature (° C)-40
  • Max Temperature (° C)121
  • Max Operating Pressure (PSI)220
  • Quantity5
  • DescriptionCompression to Threaded Fitting, Straight Adapter, PVDF, 1/4" OD x 1/4" NPT(M); 5/Pk