Compact Dehumidifiers
Convenient moisture removal in a portable design
  • Low temperature operation down to 41° F
  • 3 Fan speeds
  • Loss of power protection with auto restart
  • Automatic shut-off with bucket full indicator and alarm
  • Automatic defrost
  • Automatic Shut-off timer
  • Garden hose connector for continuous drainage
These dehumidifiers feature three humidity-level settings, three fan-speed settings, continuous dehumidification, full-tank indicator, and a water drain for continuous operation. A magnetic float within the water tank provides accurate water level readings and prevents tank overflow. When full, the tank indicator light illuminates and the unit automatically shuts off. Selectable humidity levels offer options for any comfort level. Once the selected humidity level is achieved, the compressor shuts off automatically. In situations where constant dehumidification is necessary, the continuous operation mode overrides the level setting. For convenient portability, the units feature a carrying handle and casters.
69,100.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Display Type Power (VAC) Description
Availability Pricing
Soleus DSJ-50E-01 Compact Digital Dehumidifier, 50 Pints; 120 V, 60 Hz 3770111 Mfr # DSJ-50E-01
COMPARE
115 Compact Digital Dehumidifier, 50 Pints; 120 V, 60 Hz
₹69,100.00INR / Each