Cole-Parmer® Wide-Mouth Plastic Bottles, FEP
Maintain sterility of samples
  • Inert, FEP bottles withstand repeated sterilization
  • Use in broad temperature range –328 to 401°F (–200 to 205°C)
  • Leakproof ETFE screw closure
  • Autoclavable
The most inert bottles available are those made of FEP. They will withstand repeated sterilization and temperatures from –328 to 401°F (–200 to 205°C). Use them for cryogenic experiments, trace metal analysis, and organic solvents. Has leakproof ETFE screw closure.
67,700.00 - ₹121,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
250 GL45 Fluorinated Ethylene Propylene (FEP)
₹67,700.00INR / Each
COMPARE
500 GL45 Fluorinated Ethylene Propylene (FEP)
₹84,200.00INR / Each
COMPARE
1000 GL45 Fluorinated Ethylene Propylene (FEP)
₹121,000.00INR / Each