Cole-Parmer® Wide-Form Porcelain Crucibles with Covers
Withstand temperatures up to 1150 °C (2102 °F)
  • Resistance to acids and alkalies
  • Autoclavable
These wide mouth porcelain crucibles offer easy sample placement. They are glazed on inner and outer surfaces, except outer bottom and rim. Withstand temperatures to 1150 °C (2102 °F) and heating rates up to 200 °C per hour. Offers excellent resistance to acids and alkalies, except for hydrofluoric acid.
6,200.00 - ₹19,100.00INR / Pkg of 12
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
20 10 32
₹11,900.00INR / Pkg of 12
COMPARE
26 20 41
₹12,400.00INR / Pkg of 12
COMPARE
20 10 32
₹6,200.00INR / Pkg of 6
COMPARE
26 20 41
₹6,600.00INR / Pkg of 6
COMPARE
31 30 50
₹9,700.00INR / Pkg of 6
COMPARE
37 50 61
₹16,800.00INR / Pkg of 6
COMPARE
46 100 76
₹13,900.00INR / Pkg of 6
COMPARE
52 150 84
₹16,100.00INR / Pkg of 6
COMPARE
60 250 102
₹19,100.00INR / Pkg of 6
Showing 1- 9 of 9