Cole-Parmer® Volmetric Glassware Starter Kits

No need to order multiple items!
  • Get up and running quickly with the most commonly used glassware
Convenient glassware starter kit will take care of your titration, filtration, and liquid transfer research project needs. Class B borosilicate glass has a low coefficient of expansion, for resistance to heat, and a very high resistance to chemical attack.

These kits include:
1 each of 50-, 100-, 250, 600, and 1000-mL beakers
One 9" long beaker tongs
1 each of 50-, 100-, 250, 600, and 1000-mL Erlenmeyer flasks
1 each of 10-, 25-, and 100-mL hexagonal base cylinders
1 each of 100- and 500-mL media bottles
1 two-tierd, 12 hole, test tube rack
Twelve 15 mL test tubes with rubber stoppers
One 5" long test tube clamp with finger grip
Twelve 8" long glass rods with rounded edges
Twelve 30 mL dropping bottles
57,900.00 - ₹63,300.00INR / Set of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Description
Availability Pricing
COMPARE
Glass Volumetric Glassware Starter Kit, 9 pieces
₹57,900.00INR / Set of 1
COMPARE
Glass Volumetric Glassware Starter Kit, 9 pieces, with tote
₹63,300.00INR / Set of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat