Cole-Parmer® Very-Low Differential Pressure Transducers
Uni- and bi-directional pressure measurement offer high accuracy for building energy management
  • Static accuracy of ±1%
  • Fully protected against reverse wiring
  • Fire retardent case is UL 94 V-0 approved
Low-pressure transucers sense gauge or differential pressure and convert to a proportional output. Utilizing an improved stainless steel micro-tig welded sensor, positive pressure moves the diaphragm toward the electrode, increasing capacitance. A decrease in pressure moves the diaphragm away from the electrode, decreasing the capcitance. Change in capacitance is converted to a linear DC electrical signal through the transducer''s circuit.
Transducers are designed for HVAC, air volume and fan control, static duct and clean-room pressures, and environmental pollution control.
68,000.00 - ₹153,000.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (in WC) Min Pressure (in WC) Output
Availability Pricing
COMPARE
0.5 0 4 to 20 mA
₹70,800.00INR / Each
COMPARE
1 0 4 to 20 mA
₹70,800.00INR / Each
COMPARE
5 0 4 to 20 mA
₹74,700.00INR / Each
COMPARE
10 0 4 to 20 mA
₹70,800.00INR / Each
COMPARE
25 0 4 to 20 mA
₹70,800.00INR / Each
COMPARE
50 0 4 to 20 mA
₹70,800.00INR / Each
COMPARE
100 0 4 to 20 mA
₹70,800.00INR / Each
COMPARE
0.25 0 0 to 5 VDC
₹68,000.00INR / Each
COMPARE
0.5 0 0 to 5 VDC
₹76,500.00INR / Each
COMPARE
5 0 0 to 5 VDC
₹70,800.00INR / Each
COMPARE
10 0 0 to 5 VDC
₹70,800.00INR / Each
COMPARE
50 0 0 to 5 VDC
₹70,800.00INR / Each
COMPARE
100 0 0 to 5 VDC
₹70,800.00INR / Each
COMPARE
0.25 -0.25 4 to 20 mA
₹74,600.00INR / Each
COMPARE
2.5 -2.5 4 to 20 mA
₹76,500.00INR / Each
COMPARE
50 -50 4 to 20 mA
₹151,000.00INR / Each
COMPARE
0.1 -0.1 1 to 5 VDC
₹70,800.00INR / Each
COMPARE
0.5 -0.5 1 to 5 VDC
₹70,800.00INR / Each
COMPARE
1 -1 1 to 5 VDC
₹70,800.00INR / Each
COMPARE
50 -50 1 to 5 VDC
₹153,000.00INR / Each
Showing 1- 20 of 20