Cole-Parmer® Vertical/Chest Freezer Racks with Spring Clips for Standard 2" Boxes
Spring clips securely hold boxes in rack yet allow for quick and easy removal of boxes
  • Use with a wide variety of chest freezers and liquid nitrogen tanks
  • Accommodates standard 2" cardboard, plastic, and metal boxes
  • Dual stainless steel handles for easy handling
  • Sturdy corrosion-resistant stainless steel construction
Racks feature sturdy handles for easy access to your samples in vertical/chest freezers.
22,700.00 - ₹32,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
10 boxes 22
₹24,500.00INR / Each
COMPARE
11 boxes 24 3/16
₹22,700.00INR / Each
COMPARE
13 boxes 28 1/4
₹27,300.00INR / Each
COMPARE
14 boxes 30 13/16
₹32,900.00INR / Each