Cole-Parmer® Vacuum-Driven Filtration
Ideal for large volume sample separation and purification
  • Engraved graduation ensure veracity
  • Hose connector design can fit multiple hose diameters
  • Non-pyrogenic
  • Packaged in easy peel-to-open plastic bag, and receiver bottle cap is individually wrapped
  • Each unit is individually labelled for lot traceability
  • Gamma sterilized
Vacuum filters are available in PVDF, PES and MCE with 2 different pore sizes - 0.22μm and 0.45μm. These are light weight with thick wall construction and have ergonomically designed sidewalls and collar to simplify assembling. The bottle has wide bottle mouth for efficient and stable pour out.
9,300.00 - ₹23,300.00INR / Case of 1
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem Availability Pricing
COMPARE
₹9,300.00INR / Case of 1
COMPARE
₹10,500.00INR / Case of 1
COMPARE
₹12,900.00INR / Case of 1
COMPARE
₹21,100.00INR / Case of 1
COMPARE
₹9,300.00INR / Case of 1
COMPARE
₹10,500.00INR / Case of 1
COMPARE
₹12,900.00INR / Case of 1
COMPARE
₹21,100.00INR / Case of 1
COMPARE
₹10,300.00INR / Case of 1
COMPARE
₹10,300.00INR / Case of 1
COMPARE
₹11,600.00INR / Case of 1
COMPARE
₹11,600.00INR / Case of 1
COMPARE
₹14,600.00INR / Case of 1
COMPARE
₹14,600.00INR / Case of 1
COMPARE
₹23,300.00INR / Case of 1
COMPARE
₹23,300.00INR / Case of 1
COMPARE
₹9,300.00INR / Case of 1
COMPARE
₹9,300.00INR / Case of 1
COMPARE
₹10,500.00INR / Case of 1
COMPARE
₹10,500.00INR / Case of 1
Showing 1 - 20 of 24