Cole-Parmer VAC FILTERPVDF S 150ML .22UM 12/CS
Representative image only.
Cole-Parmer VAC FILTERPVDF S 150ML .22UM 12/CS
Cole-Parmer | Item: 1597159
Ideal for large volume sample separation and purification
₹9,300.00INR / Case of 1
₹9,300.00INR / Case of 1
Product Description
Key Features
  • Engraved graduation ensure veracity
  • Hose connector design can fit multiple hose diameters
  • Non-pyrogenic
  • Packaged in easy peel-to-open plastic bag, and receiver bottle cap is individually wrapped
  • Each unit is individually labelled for lot traceability
  • Gamma sterilized
More About this Item
Vacuum filters are available in PVDF, PES and MCE with 2 different pore sizes - 0.22μm and 0.45μm. These are light weight with thick wall construction and have ergonomically designed sidewalls and collar to simplify assembling. The bottle has wide bottle mouth for efficient and stable pour out.
Specifications
  • DescriptionVAC FILTERPVDF S 150ML .22UM 12/CS