Cole-Parmer® Vacuum Desiccators with Clear Base, PC
Holds vacuum up to 29” Hg for 24 hours
  • Knurled knob provides easy handling of dome
  • Autoclavable
These vacuum desiccators can hold a vacuum up to 29” (740 mm) Hg for 24 hours without any greasing. The top dome with molded-in knurled knob is constructed of rigid, transparent polycarbonate (PC), providing you a clear view of your sample.
The lower half is made of transparent PC for increased visibility and includes a perforated polypropylene (PP) desiccator plate. An internal groove on the flange holds the silicon rubber O-ring that provides leakproof vacuum capability.
A PP stopcock accepts ¼” (6.4 mm) ID vacuum tubing and features a self-lubricating PTFE plug. The three-way plug allows you to create, hold, and release the vacuum. Autoclavable at 120°C for 15 minutes.
21,000.00 - ₹73,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clearance Above Plate (mm) Flange Outside Diameter (OD) (cm) Material
Availability Pricing
COMPARE
123 17.1 Polycarbonate
₹21,000.00INR / Each
COMPARE
166 23.2 Polycarbonate
₹33,800.00INR / Each
COMPARE
207 27.6 Polycarbonate
₹43,400.00INR / Each
COMPARE
255 34 Polycarbonate
₹73,200.00INR / Each