Cole-Parmer® Upright Freezer Racks for Standard 3.75" High Boxes
Get organized for easier retrieval of samples
  • Accommodates stardard 3.75" high polycarbonate boxes
  • Easily access boxes from either side of rack
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
14,300.00 - ₹33,800.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 2 Array 8 1/16 16 5/16
₹14,300.00INR / Each
COMPARE
3 x 3 Array 12 1/16 16 5/16
₹21,500.00INR / Each
COMPARE
3 x 4 Array 16 1/8 16 5/16
₹28,800.00INR / Each
COMPARE
4 x 2 Array 8 1/16 21 15/16
₹18,600.00INR / Each
COMPARE
4 x 3 Array 12 1/16 21 15/16
₹27,000.00INR / Each
COMPARE
5 x 2 Array 8 1/16 26 3/4
₹24,600.00INR / Each
COMPARE
5 x 3 Array 12 1/16 26 3/4
₹33,800.00INR / Each
Showing 1- 7 of 7