Cole-Parmer® Upright Freezer Drawer Racks for Microtube Boxes with 2 7/16" Max Height
Drawers slide out independently for quick access and storage of boxes
  • Easily access boxes from either side of rack
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer.
30,700.00 - ₹66,200.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 3 Array 8 10 3/4
₹30,700.00INR / Each
COMPARE
4 x 3 Array 8 14 3/16
₹35,000.00INR / Each
COMPARE
4 x 4 Array 10 5/8 14 3/16
₹43,500.00INR / Each
COMPARE
4 x 5 Array 13 3/16 14 3/16
₹52,200.00INR / Each
COMPARE
5 x 3 Array 8 17 3/4
₹42,700.00INR / Each
COMPARE
5 x 4 Array 10 5/8 17 3/4
₹53,700.00INR / Each
COMPARE
5 x 5 Array 13 3/16 17 3/4
₹58,600.00INR / Each
COMPARE
6 x 3 Array 8 21 3/16
₹50,600.00INR / Each
COMPARE
6 x 4 Array 10 5/8 21 3/16
₹57,600.00INR / Each
COMPARE
6 x 5 Array 13 3/16 21 3/16
₹66,200.00INR / Each
Showing 1- 10 of 10