Cole-Parmer® Tube Sinkers, PVDF (Kynar®)
Weighted sinkers help keep tubing at the bottom of container
  • Flow-through design—perfect for filling and dispensing
  • PDVF (Kynar) construction offers superior chemical resistance
  • Hose barb fitting accepts flexible tubing
  • ASME-BPE, USP Class VI, FDA 21 CFR 177.2510, REACH, and RoHS compliant
  • Animal-derived ingredient free (ADIF) and animal-derived component free (ADCF)
  • Sterilize by autoclave, gamma irradiation, x-ray, or EtO
Connect a tube sinker to your tubing to keep it at the bottom of your container. The flow-through design provides efficient emptying or filling by pumps or dispensers. Slight recess at the bottom of the sinker prevents it from suctioning against the container. Monobarb fitting accepts flexible tubing.
6,500.00 - ₹19,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in)
Availability Pricing
COMPARE
3/16
₹6,500.00INR / Each
COMPARE
1/4
₹6,600.00INR / Each
COMPARE
5/16
₹7,000.00INR / Each
COMPARE
3/8
₹12,600.00INR / Each
COMPARE
1/2
₹12,200.00INR / Each
COMPARE
3/4
₹19,400.00INR / Each